Learn More
anti-GRHPR Matched Antibody Pair, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-GRHPR (100μg); Mouse monoclonal anti-GRHPR, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes an enzyme with hydroxypyruvate reductase, glyoxylate reductase, and D-glycerate dehydrogenase enzymatic activities. The enzyme has widespread tissue expression and has a role in metabolism. Type II hyperoxaluria is caused by mutations in this gene.
ELISA
Specifications
Specifications
| Antigen | GRHPR |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GRHPR. |
| Gene | GRHPR |
| Gene Alias | GLXR/GLYD/PH2 |
| Gene Symbols | GRHPR |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.