Learn More
FTL, Mouse, Clone: 1C2, Abnova™
Mouse monoclonal antibody raised against a full-length native FTL.
Brand: Abnova H00002512-M07.100ug
Description
This gene encodes the light subunit of the ferritin protein. Ferritin is the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in this light chain ferritin gene are associated with several neurodegenerative diseases and hyperferritinemia-cataract syndrome. This gene has multiple pseudogenes. [provided by RefSeq
Specifications
| FTL | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| FTL | |
| MGC71996 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2512 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ELISA, Immunofluorescence | |
| 1C2 | |
| Mouse monoclonal antibody raised against a full length native FTL. | |
| PBS with no preservative; pH 7.4 | |
| P02792 | |
| FTL | |
| Native purified human FTL. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.