missing translation for 'onlineSavingsMsg'
Learn More

Anti-FGL1 antibody [RM1297], Abcam

Product Code. 32073854 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
1 mL
Unit Size:
100µL
1mL
20µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32073854 1 mL 1mL
32074685 100 μL 100µL
32073412 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32073854 Supplier Abcam Supplier No. ab3236111mL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Polyclonal Antibody

Rabbit Recombinant Multiclonal FGL1 antibody. Suitable for WB, IHC-P, ICC/IF, Flow Cyt (Intra), IP and reacts with Human samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen FGL1
Applications Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Immunoprecipitation, Western Blot
Cellular Localization Secreted; Secreted in the blood plasma.
Classification Recombinant Polyclonal
Clone RM1297
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene FGL1
Gene Accession No. Q08830
Host Species Rabbit
Immunogen FGL1
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immuno-oncology
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene FGL1
Target Molecule FGL1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.