missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Anti-Ferritin Antibody [RM1036], Rabbit Multiclonal, 100 μL
Description
- Host Species: Rabbit
- Clonality: Multiclonal
- Isotype: IgG
- Applications: WB, IHC-P, Flow Cyt (Intra)
- Species Reactivity: Human, Mouse, Rat
- Form: Liquid
- Storage Buffer: PBS, 40% Glycerol, 0.05% BSA, 0.01% Sodium azide, pH 7.2 to 7.4
- Citations: 8 publications
Specifications
Specifications
| Buffer | PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH7.2 to 7.4 |
| Detection Location | In-Blot Detection |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Target Molecule | Ferritin Heavy Chain (Human, Mouse, Rat) |
| Cross Reactivity | Mouse, Human, Rat |
| Formulation | PBS, 40% glycerol, 0.05% BSA, 0.01% sodium azide, pH 7.2 to 7.4 |
| Quantity | 100 μL |
| For Use With (Application) | Flow Cytometry (Intra), Western Blot, IHC-P |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?