Learn More
F3, Human, IP-WB Antibody Pair (PW3), Abnova™
Product Code. 16112014
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16112014 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16112014 Supplier Abnova Supplier No. H00002152PW3.1Set
 Restricted Item
Estimated Shipment: 06-10-2026
Includes
Antibody pair for IP: rabbit polyclonal anti-F3 (300μL), Antibody pair for WB: mouse purified polyclonal anti-F3 (50μg)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen F3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene F3
Gene Alias CD142/TF/TFA
Gene Symbols F3
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2152
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-F3 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-F3 (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid