missing translation for 'onlineSavingsMsg'
Learn More

Anti-ERK2 antibody, Abcam

Product Code. 32055917 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Unit Size:
1mL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32055917 1 mL 1mL
32053937 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32055917 Supplier Abcam Supplier No. ab152821mL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Anti-ERK2 antibody

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Mapk1 (ERK2)
Applications Immunohistochemistry (FFPE)
Cellular Localization Cytoplasm; Cytoskeleton; Spindle; Nucleus; Cytoplasm; Cytoskeleton; Microtubule organizing center; Centrosome; Cytoplasm; Membrane; Caveola; Cell junction; Focal adhesion; Associated with the spindle during prometaphase and metaphase (By similarity). PEA15-binding and phosphorylated DAPK1 promote its cytoplasmic retention. Phosphorylation at Ser-244 and Ser-246 as well as autophosphorylation at Thr-188 promote nuclear localization (By similarity). Localization to focal adhesions is stimulated by EGF (PubMed:15923189).
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.6, Preservative: 0.1% Sodium azide, Constituents: PBS, 1% BSA
Gene Mapk1
Gene Accession No. P63085
Host Species Rabbit
Immunogen Mapk1 (ERK2)
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Oncology
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene Mapk1
Target Molecule Mapk1 (ERK2)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.