Learn More
anti-ERCC6L, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00054821-PW1.1Set
Product Code. 16133454
  • 843.00€ / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-ERCC6L (300μL), Antibody pair for WB: mouse purified polyclonal anti-ERCC6L (50μg)
Specifications
ERCC6L
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
FLJ20105/MGC131695/PICH
Mouse, Rabbit
RUO
54821
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
ERCC6L
ERCC6L
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-ERCC6L (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-ERCC6L (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.