Learn More
anti-EPCAM, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00004072-PW2.1Set
Product Code. 16172304
  • 843.00€ / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-EPCAM (300μL), Antibody pair for WB: mouse purified polyclonal anti-EPCAM (50μg)
Specifications
EPCAM
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
17-1A/323/A3/CD326/CO-17A/CO17-1A/EGP/EGP-2/EGP34/EGP40/ESA/Ep-CAM/GA733-2/HEA125/KS1/4/KSA/M4S1/MH99/MIC18/MK-1/MOC31/TACST-1/TACSTD1/TROP1/hEGP-2
Mouse, Rabbit
RUO
4072
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
EPCAM
EPCAM
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-EPCAM (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-EPCAM (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.