Learn More
elastin, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Description
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal dominant cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | elastin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human ELN protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ELN |
| Gene Accession No. | BC065566.1 |
| Gene Alias | FLJ38671/FLJ43523/SVAS/WBS/WS |
| Gene Symbols | ELN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.