Learn More
anti-EFHD1, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00080303-PW1.1Set
Product Code. 16153714
  • 843.00€ / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-EFHD1 (300μL), Antibody pair for WB: mouse polyclonal anti-EFHD1 (50μL)
Specifications
EFHD1
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
DKFZp781H0842/FLJ13612/MST133/MSTP133/PP3051
Mouse, Rabbit
RUO
80303
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
EFHD1
EFHD1
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-EFHD1 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-EFHD1 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.