missing translation for 'onlineSavingsMsg'
Learn More

Dynamin II Mouse, Unlabeled, Clone: 27, BD

Product Code. 15855369
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15855369 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15855369 Supplier BD Biosciences Supplier No. 610263
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dynamin is a membrane-associated GTPase that binds to GTP, microtubules, and phospholipids. Northern blot analysis has shown dynamin to be present in most tissues but at an amount about 20-fold lower than that in brain. There are at least two distinct dynamin genes in mammals. Transcripts of both dynamin genes are alternatively spliced at two or more sites. The first site is identical in bothdynamins, whereas, the second site differs. The two gene products are known as Dynamin I and Dynamin II and show 79% identity. Dynamin I is expressed almost exclusively in the central nervous system while Dynamin II expression is ubiquitous. The two proteins are highly homologous in the N-terminal region, while the C-terminal domain shows significant divergence. The GTPase activity of Dynamin I is stimulated several fold by binding to microtubules, phospholipids, and membranous vesicles. Dynamin I is a good substrate of PKC in vitro as well as in vivo in resting nerve terminals. In vitro, phosphorylation occurs at the C-terminus of Dynamin I and this enhances the GTPase activity more than 10-fold. However, Dynamin II is not a substrate of PKC and its activity does not appear to be affected by phosphorylation. These data suggest that the function of the common N-terminal domain between Dynamin I and II may be differentially specified by distinct C-terminal domains.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Dynamin II
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 27
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.