Learn More
anti-DNAI1, Human, Matched Antibody Pair, Abnova™
Product Code. 16133324
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16133324 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16133324 Supplier Abnova Supplier No. H00027019AP22.1Set
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Includes
Capture antibody: rabbit MaxPab affinity purified polyclonal anti-DNAI1 (100μg) , Detection antibody: mouse purified polyclonal anti-DNAI1 (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen DNAI1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human DNAI1.
Gene DNAI1
Gene Alias CILD1/ICS/ICS1/MGC26204/PCD
Gene Symbols DNAI1
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27019
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-DNAI1 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-DNAI1 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Mostrar más Mostrar menos
For Research Use Only
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.