missing translation for 'onlineSavingsMsg'
Learn More

Anti-CXCL13 antibody [RM1170], Abcam

Product Code. 32070663 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
1 mL
Unit Size:
100µL
1mL
20µL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32070663 1 mL 1mL
32070356 100 μL 100µL
32071284 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 32070663 Supplier Abcam Supplier No. ab3175081mL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Polyclonal Antibody

Rabbit Recombinant Multiclonal BCA1 antibody. Suitable for IHC-P and reacts with Human, Mouse samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen CXCL13 (BCA1)
Applications Immunohistochemistry (FFPE)
Cellular Localization Secreted
Classification Recombinant Polyclonal
Clone RM1170
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene CXCL13
Gene Accession No. O43927
Host Species Rabbit
Immunogen CXCL13 (BCA1)
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene CXCL13
Target Molecule CXCL13 (BCA1)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.