Learn More
CtBP2 (C-Terminal Binding Protein-2) Mouse, Unlabeled, Clone: 16, BD
Description
Transcriptional repression is critical for genetic regulation of many cellular proteins, and involves either a transcription regulator that has an intrinsic repressor domain, or a transcription regulator that contains a binding site in the regulatory domain where a corepressor can bind. C-terminal binding proteins (CtBP1 and CtBP2) are a class of corepressors that interact with a subset of transcription factors through a PLDLSL sequence motif. CtBP1 and CtBP2 are 80% homologous, and both are expressed at high levels in embryonic tissues. However, CtBP1 is expressed higher in adult tissues, and is more widely expressed in embryonic and adult tissues. Both CtBP1 and CtBP2 bind the δEF1 zinc finger-homeodomain transcription factor, and enhance transcriptional repression via interaction with the PLDLSL-sequence in δEF1. CtBP1 and CtBP2 bind other zinc finger transcription factors, including Kheper and BKLF. In addition, CtBP2 interacts with other transcription factors, such as hFOG-2, Evi-1, AREB6, and ZEB. Thus, CtBP1 and CtBP2 may be important corepressors for a variety of transcriptional factors.
Host Species: Mouse
Clone: 16
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse CtBP2 aa. 361-445
Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | CtBP2 (C-Terminal Binding Protein-2) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 16 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Description | C-Terminal Binding Protein-2 |
| Formulation | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Mouse CtBP2 aa. 361-445 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.