Learn More
anti-COBRA1 Matched Antibody Pair, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-COBRA1 (100μg); Mouse purified polyclonal anti-COBRA1 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2; MIM 606026), either NELFC or NELFD (TH1L; MIM 605297), and NELFE (RDBP; MIM 154040). NELF acts with DRB sensitivity-inducing factor (DSIF), a heterodimer of SPT4 (SUPT4H1; MIM 603555) and SPT5 (SUPT5H; MIM 602102), to cause transcriptional pausing of RNA polymerase II.
ELISA
Specifications
Specifications
| Antigen | COBRA1 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human COBRA1. |
| Gene | COBRA1 |
| Gene Alias | DKFZp586B0519/KIAA1182/NELF-B/NELFB |
| Gene Symbols | COBRA1 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.