missing translation for 'onlineSavingsMsg'
Learn More

connector enhancer of kinase suppressor of Ras 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16152246
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16152246 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16152246 Supplier Abnova Supplier No. H00010256A01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a partial recombinant CNKSR1.

This gene is a necessary element in receptor tyrosine kinase pathways, possibly as a tyrosine phosphorylation target. It is involved in regulation of RAF in the MAPK pathway and may also play a role in a MAPK-independent pathway. [provided by RefSeq

Sequence: VLCAAVELLHEADALLFWLSRYLFSHLNDFSACQEIRDLLEELSQVLHEDGPAAEKEGTVLRICSHVAGICHNILVCCPKELLEQKAVLEQVQLDSPLGL

Specifications

Antigen connector enhancer of kinase suppressor of Ras 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CNKSR1.
Formulation 50% glycerol
Gene CNKSR1
Gene Accession No. NM_006314
Gene Alias CNK/CNK1/KSR
Gene Symbols CNKSR1
Host Species Mouse
Immunogen CNKSR1 (NP_006305, 111 a.a. ∼ 210 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10256
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.