missing translation for 'onlineSavingsMsg'
Learn More

Anti-CLPP antibody [EPR7133], Abcam

Product Code. 32075007 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32075007 100 μL 100µL
32073757 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32075007 Supplier Abcam Supplier No. ab124822100uL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Rabbit Recombinant Monoclonal CLPP antibody. Suitable for IHC-P, IP, WB and reacts with Mouse, Rat, Human samples. Cited in 52 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen CLPP
Applications Immunohistochemistry (FFPE), Immunoprecipitation, Western Blot
Cellular Localization Mitochondrion matrix
Classification Recombinant Monoclonal
Clone EPR7133
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene CLPP
Gene Accession No. Q16740
Host Species Rabbit
Immunogen CLPP
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Research Discipline Unassigned
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Conditional Ambient
Target Gene CLPP
Target Molecule CLPP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.