Learn More
CD73 Mouse anti-Rat, Unlabeled, Clone: 5F/B9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551123
Description
The 5F/B9 antibody reacts with CD73 or Ecto-5'-nucleotidase (5'-NT), a 68-kDa GPI-anchored cell-surface protein with enzymatic and signal transduction activities. 5'-NT catalyzes the dephosphorylation of extracellular nucleoside 5' monophosphates into a form which can enter cells to meet their metabolic needs. It also regulates the concentration of extracellular adenosine, which initiates a variety of physiological responses through the adenosine receptors in many tissues. Membrane-bound and/or intracytoplasmic CD73 is found in a variety of organs and tissues, including liver, kidney, adrenal cortex, spleen, muscle, nervous tissue, and heart. The expression of CD73 on mouse leukocytes has been well characterized. In the rat, CD73 has been detected on subsets of peripheral B and T lymphocytes, NK cells, and granulocytes, as well as bone marrow myeloid cells and some CD4+CD8- thymocytes. The 5F/B9 mAb does not inhibit the enzymatic activity of 5’-NT. Although soluble or plate-bound 5F/B9 mAb by itself does not affect T lymphocyte proliferation, it is an effective co-stimulator with plate-bound anti-rat CD3 mAb G4.18 (Cat. no. 554829).
Specifications
| CD73 | |
| Monoclonal | |
| 0.5mg/mL | |
| Ecto-5'-nucleotidase | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 5F/B9 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Ecto-5′-nucleotidase purified from rat renal membranes | |
| 0.1 mg | |
| Immunology | |
| Rat | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.