missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD4; Clone: L200; BD
Description
Host Species: Mouse
Clone L200
Isotype IgG1, κ
Species Reactivity Hu, Rh, Cyn, Bb
Flow Cytometry
Specifications
Specifications
| Antigen | CD4 |
| Classification | Monoclonal |
| Clone | L200 |
| Conjugate | Brilliant Violet 421 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Baboon, Cynomolgus, Human, Rhesus Monkey |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?