missing translation for 'onlineSavingsMsg'
Learn More

Anti-CD4 antibody, Abcam

Product Code. 32057363 Shop All Abcam Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
Unit Size:
100µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32057363 500 μL 500µL
32056106 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32057363 Supplier Abcam Supplier No. ab231460500uL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Anti-CD4 antibody

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen CD4
Applications Immunohistochemistry (FFPE)
Cellular Localization Cell membrane; Single-pass type I membrane protein; Localizes to lipid rafts (PubMed:12517957, PubMed:9168119). Removed from plasma membrane by HIV-1 Nef protein that increases clathrin-dependent endocytosis of this antigen to target it to lysosomal degradation. Cell surface expression is also down-modulated by HIV-1 Envelope polyprotein gp160 that interacts with, and sequesters CD4 in the endoplasmic reticulum.
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.6, Preservative: 0.1% Sodium azide, Constituents: PBS, 1% BSA
Gene CD4
Gene Accession No. P01730
Host Species Rabbit
Immunogen CD4
Purification Method Affinity purification
Quantity 500 μL
Regulatory Status RUO
Research Discipline Research Tools
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene CD4
Target Molecule CD4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.