missing translation for 'onlineSavingsMsg'
Learn More

Anti-CD38 antibody [90], Abcam

Product Code. 32047300 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 μg
20 μg
Unit Size:
10µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32047300 10 μg 10µg
32045904 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32047300 Supplier Abcam Supplier No. ab6140010ug
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody

Rat Monoclonal CD38 antibody. Suitable for Flow Cyt, IHC-Fr and reacts with Mouse samples. Cited in 1 publication.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Cd38
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Cellular Localization Membrane; Single-pass type II membrane protein
Classification Monoclonal
Clone '90
Conjugate Unconjugated
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS
Gene Cd38
Gene Accession No. P56528
Host Species Rat
Immunogen Cd38
Purification Method Affinity purification
Quantity 10 μg
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG2a
Shipping Condition Blue Ice
Target Gene Cd38
Target Molecule Cd38
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.