missing translation for 'onlineSavingsMsg'
Learn More

CD32 Mouse anti-Human, Unlabeled, Clone: FLI8.26, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 557333

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15898878

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »


Description

Description

Reacts with the 40kDa type I membrane glycoprotein, FcγRII, the low affinity receptor for aggregated IgG expressed on B cells, monocytes, granulocytes, and platelets. CD32 mediates endocytosis, cytotoxicity, platelet activation and immunomodulation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: FLI8.26 (2003)
Isotype: IgG2b, κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus

Flow Cytometry

Specifications

Specifications

CD32
Monoclonal
0.5mg/mL
CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Flow Cytometry
FLI8.26
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
K562 or FcγRII+ L cell lines
0.1 mg
Immunology
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.