Learn More
CD32 Mouse anti-Human, Unlabeled, Clone: FLI8.26, BD
Mouse Monoclonal Antibody
208.00€ - 331.00€
Specifications
Antigen | CD32 |
---|---|
Clone | FLI8.26 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15818178
|
BD Biosciences
555447 |
0.1 mg |
208.00€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
15898878
|
BD Biosciences
557333 |
0.1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
Reacts with the 40kDa type I membrane glycoprotein, FcγRII, the low affinity receptor for aggregated IgG expressed on B cells, monocytes, granulocytes, and platelets. CD32 mediates endocytosis, cytotoxicity, platelet activation and immunomodulation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: FLI8.26 (2003)
Isotype: IgG2b, κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Flow Cytometry
Specifications
CD32 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C |
FLI8.26 | |
Flow Cytometry | |
Unconjugated | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
K562 or FcγRII+ L cell lines | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.