Learn More
CD32 Mouse anti-Human, Unlabeled, Clone: FLI8.26, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555447
Additional Details : Weight : 0.01000kg
Description
Reacts with the 40kDa type I membrane glycoprotein, FcγRII, the low affinity receptor for aggregated IgG expressed on B cells, monocytes, granulocytes, and platelets. CD32 mediates endocytosis, cytotoxicity, platelet activation and immunomodulation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: FLI8.26 (2003)
Isotype: IgG2b, κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Flow Cytometry
Specifications
CD32 | |
Monoclonal | |
0.5mg/mL | |
CD32a,FcγRIIa, FCGR2A; CD32b, FcγRIIb, FCGR2B; CD32c, FcγRIIc, FCGR2C | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
FLI8.26 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
K562 or FcγRII+ L cell lines | |
0.1 mg | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG2b κ |
For Research Use Only.