missing translation for 'onlineSavingsMsg'
Learn More

Anti-CD31 antibody, Abcam

Product Code. 32107019 Shop All Abcam Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
Unit Size:
1mL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32107019 1 mL 1mL
32097475 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32107019 Supplier Abcam Supplier No. ab283641mL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rabbit Polyclonal CD31 antibody. Suitable for IHC-P and reacts with Human samples. Cited in 2751 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen PECAM1 (CD31)
Applications Immunohistochemistry (FFPE)
Cellular Localization Cell membrane; Single-pass type I membrane protein; Cell surface expression on neutrophils is down-regulated upon fMLP or CXCL8/IL8-mediated stimulation.; Isoform Long: Cell membrane; Single-pass type I membrane protein; Membrane raft; Cell junction; Localizes to the lateral border recycling compartment (LBRC) and recycles from the LBRC to the junction in resting endothelial cells.; Isoform Delta15: Cell junction; Localizes to the lateral border recycling compartment (LBRC) and recycles from the LBRC to the junction in resting endothelial cells.
Classification Polyclonal
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.1% Sodium azide, Constituents: PBS, 1% BSA
Gene PECAM1
Gene Accession No. P16284
Host Species Rabbit
Immunogen PECAM1 (CD31)
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immuno-oncology
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene PECAM1
Target Molecule PECAM1 (CD31)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.