missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3, Mouse anti-Human, Clone: SK7, BV421, BD
Brand: BD Biosciences 563797
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: SK7
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human Thymocytes
Flow cytometry
Specifications
| CD3 | |
| SK7 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| Brilliant Violet 421 | |
| Human Thymocytes | |
| 25 Tests | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction