Learn More
CD124 Mouse anti-Human, PE, Clone: HIL4R-M57, BD
Description
The hIL4R-M57 antibody specifically binds to the α subunit (IL-4R α ) of the human Interleukin-4 Receptor complex which is also known as CD124. The human IL-4R α , also known as B cell stimulatory factor 1 receptor (BSF-1 receptor), is a 140 kDa transmembrane glycoprotein that is expressed by B and T lymphocytes and a variety of other hematopoietic and non-hematopoietic cells and cell lines. The cell surface IL-4R α chain binds IL-4 with high affinity and associates with either the common γchain (IL-4R α / γα exists in soluble form in biological fluids. In contrast to mice, in humans no distinct mRNA coding for sIL-4R has been described, suggested that human sIL4-R is exclusively produced by proteolytic cleavage of the cell surface receptor. The serum levels of soluble IL-4R α appear to elevate in pathological situations such as allergy and parasitic infections. Depending on the ratios of IL-4 and sIL-4R α present in the local milieu, the sIL-4R α may augment or antagonize the activities of IL-4. The immunogen used to generate the hIL4R-M57 hybridoma was soluble human IL-4R.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
Specifications
| Antigen | CD124 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HIL4R-M57 |
| Conjugate | PE |
| Description | IL-4 Receptor α Chain, CD124 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Soluble Human IL-4 Receptor |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.