Learn More
anti-CD11c; Clone: B-ly6; BD
230.65€ - 402.48€
Specifications
| Antigen | CD11c |
|---|---|
| Clone | B-ly6 |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 395 |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15803099
|
BD Biosciences
563788 |
25 Tests |
231.00€ 230.65€ / Each Save 0.35€ 0.15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15893089
|
BD Biosciences
563787 |
100 Tests |
403.00€ 402.48€ / Each Save 0.52€ 0.13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Host Species: Mouse
Clone: B-ly6
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Spleen Cells from Human with Hairy Cell Leukemia
Flow cytometry
Specifications
| CD11c | |
| Monoclonal | |
| Mouse | |
| Human | |
| Primary |
| B-ly6 | |
| Brilliant Ultraviolet 395 | |
| RUO | |
| Spleen Cells from Human with Hairy Cell Leukemia | |
| Affinity Purified |