missing translation for 'onlineSavingsMsg'
Learn More

β-Catenin Mouse, Alexa Fluor 488, Clone: 14/Beta-Catenin, BD

Product Code. 15801989
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15801989 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15801989 Supplier BD Biosciences Supplier No. 562505
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 14/Beta-Catenin monoclonal antibody specifically binds to Beta-Catenin ( β -Catenin). β -Catenin is a 92 kDa protein that binds to the cytoplasmic tail of E-Cadherin. The cadherins, transmembrane adhesion molecules, are found with catenins at adherens junctions (zonula adherens). Deletions in the cytoplasmic domain of E-Cadherin which eliminate catenin binding also result in a loss of cell adhesion, indicating that this binding is essential for E-Cadherin function. Although the α - and β -Catenins have been cloned, very little is known about their biochemical roles. However a link between β -Catenin and colon cancer has been described. β -Catenin was found to co-immunoprecipitate with the APC tumor suppressor protein in human colorectal tumor cell lines, as well as in human kidney 293 cells. E-Cadherin, however, was not detectable in these complexes. Thus the APC-Catenin complex may be affecting the transmission of contact inhibition signals and/or the regulation of cell adhesion.

TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Catenin
Applications In vitro Assay, Immunofluorescence
Classification Monoclonal
Clone 14/Beta-Catenin
Conjugate Alexa Fluor 488
Description CTNNB1; CTNB1; Catenin beta-1; catenin (cadherin-associated protein) beta 1
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse β-Catenin aa. 571-781
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.