Learn More
capping protein (actin filament), gelsolin-like, Mouse, Clone: 6D6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CAPG.
Brand: Abnova H00000822-M02.100ug
Description
This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in nonmuscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described. [provided by RefSeq]
Sequence: AALYKVSDATGQMNLTKVADSSPFALELLISDDCFVLDNGLCGKIYIWKGRKANEKERQAALQVAEGFISRMQYAPNTQVEILPQGHESPIFKQFFKDWKSpecifications
| capping protein (actin filament), gelsolin-like | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001747 | |
| CAPG | |
| CAPG (NP_001738, 249 a.a. ∼ 348 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 822 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 6D6 | |
| Mouse monoclonal antibody raised against a partial recombinant CAPG. | |
| CAPG | |
| AFCP/MCP | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.