Learn More
C4BPB, Human, IP-WB Antibody Pair (PW1), Abnova™
Product Code. 16181804
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16181804 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16181804 Supplier Abnova Supplier No. H00000725PW1.1Set
Includes
Antibody pair for IP: mouse monoclonal anti-C4BPB (300μg), Antibody pair for WB: rabbit polyclonal anti-C4BPB (50μL)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen C4BPB
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene C4BPB
Gene Alias C4BP
Gene Symbols C4BPB
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 725
Includes Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-C4BPB (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-C4BPB (50 ul)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid