missing translation for 'onlineSavingsMsg'
Learn More

Anti-C1QA antibody [EPR2980Y], Abcam

Product Code. 32136767 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32136767 20 μL 20µL
32134692 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32136767 Supplier Abcam Supplier No. ab7642520uL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Rabbit Recombinant Monoclonal C1QA antibody. Suitable for IHC-P, WB and reacts with Human samples. Cited in 2 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen C1QA
Applications Immunohistochemistry (FFPE), Western Blot
Cellular Localization Secreted; Cell surface; Specifically binds IgG or IgM immunoglobulins complexed with antigens, forming antigen-antibody complexes on the surface of pathogens.
Classification Recombinant Monoclonal
Clone EPR2980Y
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene C1QA
Gene Accession No. P02745
Host Species Rabbit
Immunogen C1QA
Purification Method Affinity purification
Quantity 20 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Conditional Ambient
Target Gene C1QA
Target Molecule C1QA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.