Learn More
anti-BST2 Matched Antibody Pair, Ab-2, Abnova
Description
Includes
Rabbit MaxPab7asterisk; affinity purified polyclonal anti-BST2 (100μg); Mouse purified polyclonal anti-BST2 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Bone marrow stromal cells are involved in the growth and development of B-cells. The specific function of the protein encoded by the bone marrow stromal cell antigen 2 is undetermined; however, this protein may play a role in pre-B-cell growth and in rheumatoid arthritis.
ELISA
Specifications
Specifications
| Antigen | BST2 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human BST2. |
| Gene | BST2 |
| Gene Alias | CD317 |
| Gene Symbols | BST2 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.