missing translation for 'onlineSavingsMsg'
Learn More

Anti-BRN3A antibody [EP1972Y], Abcam

Product Code. 32171167 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 μL
20 μL
Unit Size:
10µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32171167 10 μL 10µL
32167777 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32171167 Supplier Abcam Supplier No. ab8121310uL
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Rabbit Recombinant Monoclonal BRN3A antibody. Suitable for WB, Flow Cyt (Intra) and reacts with Mouse, Rat, Human samples. Cited in 19 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen POU4F1 (BRN3A)
Applications Flow Cytometry (Intracellular Staining), Western Blot
Cellular Localization Nucleus; Cytoplasm
Classification Recombinant Monoclonal
Clone EP1972Y
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene POU4F1
Gene Accession No. Q01851
Host Species Rabbit
Immunogen POU4F1 (BRN3A)
Purification Method Affinity purification
Quantity 10 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Conditional Ambient
Target Gene POU4F1
Target Molecule POU4F1 (BRN3A)
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.