Learn More
anti-BLK, Human, IP-WB Antibody Pair, Abnova™
Product Code. 16003348
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16003348 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16003348 Supplier Abnova Supplier No. H00000640PW1.1Set
 Restricted Item
Estimated Shipment: 06-10-2026
Includes
Antibody pair for IP: rabbit polyclonal anti-BLK (300μL), Antibody pair for WB: mouse purified polyclonal anti-BLK (50μg)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen BLK
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene BLK
Gene Alias MGC10442
Gene Symbols BLK
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 640
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-BLK (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-BLK (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less
For Research Use Only
Product Title
Select an issue
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.