Learn More
anti-B2M, Human, Matched Antibody Pair, Abnova™
Product Code. 16111774
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16111774 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16111774 Supplier Abnova Supplier No. H00000567AP41.1Set
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
Includes
Capture antibody: mouse monoclonal anti-B2M (100μg) , Detection antibody: biotinylated mouse monoclonal anti-B2M, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen B2M
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human B2M.
Gene B2M
Gene Symbols B2M
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 567
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-B2M (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-B2M, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid