Learn More
anti-ATF4, Human, Matched Antibody Pair, Abnova™
Product Code. 16161764
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16161764 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16161764 Supplier Abnova Supplier No. H00000468AP41.1Set
Includes
Capture antibody: mouse monoclonal anti-ATF4, IgG1 Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-ATF4, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
To detect and quantify protein level of human
Specifications
Antigen ATF4
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ATF4.
Gene ATF4
Gene Alias CREB-2/CREB2/TAXREB67/TXREB
Gene Symbols ATF4
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 468
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-ATF4, IgG1 Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-ATF4, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid