Learn More
β-Arrestin Mouse, Unlabeled, Clone: 10, BD
Mouse Monoclonal Antibody
343.00€ - 675.00€
Specifications
Antigen | β-Arrestin |
---|---|
Clone | 10 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15805519
|
BD Biosciences
610550 |
50 μg |
343.00€
1 set |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
15815519
|
BD Biosciences
610551 |
150 μg |
675.00€
1 set |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
Description
β-Arrestins were discovered due to their ability to modulate interactions between the phosphorylated β2-Adrenergic receptors and G proteins. This modulation results in diminished β2-Adrenergic receptor function, also known as desensitization. Because arrestins are found at the synaptic terminals, they may provide a termination mechanism that allows the neurons to regain their original polarization and respond to a new neurotransmitter stimulus. The C-terminal region of arrestins is involved in selecting the phosphorylated and activated adrenergic receptors. The β-Arrestin1 gene encodes a protein of 418 amino acids with an approximate molecular weight of 55kDa. β-Arrestin1 protein is highly homologous to the 45kDa β-Arrestin2. Both proteins are widely expressed, but are especially abundant in the central nervous system.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
β-Arrestin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
10 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Rat β-Arrestin1 aa. 262-409 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.