Learn More
APOBEC3C, Human, IP-WB Antibody Pair (PW1), Abnova™
Product Code. 16004029
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16004029 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16004029 Supplier Abnova Supplier No. H00027350PW1.1Set
Includes
Antibody pair for IP: rabbit polyclonal anti-APOBEC3C (300μL), Antibody pair for WB: mouse purified polyclonal anti-APOBEC3C (50μg)
For immunoprecipitation and to detect the precipitated protein in western blot.
Specifications
Antigen APOBEC3C
Applications Immunoprecipitation, Western Blot
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
Gene APOBEC3C
Gene Alias APOBEC1L/ARDC2/ARDC4/ARP5/MGC19485/PBI/bK150C2.3
Gene Symbols APOBEC3C
Host Species Mouse, Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27350
Includes Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-APOBEC3C (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-APOBEC3C (50 ug)
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid