Learn More
ANXA5, Human, IP-WB Antibody Pair (PW3), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00000308-PW3.1Set
Product Code. 16121734
  • 843.00€ / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-ANXA5 (300μL), Antibody pair for WB: mouse purified polyclonal anti-ANXA5 (50μg)
Specifications
ANXA5
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
ANX5/ENX2/PP4
Mouse, Rabbit
RUO
308
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
ANXA5
ANXA5
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-ANXA5 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-ANXA5 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.