Learn More
anti-ANXA3 Matched Antibody Pair, Abnova
Product Code. 16161724
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16161724 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16161724 Supplier Abnova Supplier No. H00000306AP21.1Set
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-ANXA3 (100μg); Mouse purified polyclonal anti-ANXA3 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions in the inhibition of phopholipase A2 and cleavage of inositol 1,2-cyclic phosphate to form inositol 1-phosphate. This protein may also play a role in anti-coagulation.
ELISA
Specifications
Antigen ANXA3
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ANXA3.
Gene ANXA3
Gene Alias ANX3
Gene Symbols ANXA3
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ANXA3 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-ANXA3 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.