Learn More
ANXA3, Human, IP-WB Antibody Pair (PW1), Abnova™
Description
Includes
Antibody pair for IP: mouse monoclonal anti-ANXA3 (300μg), Antibody pair for WB: rabbit polyclonal anti-ANXA3 (50μL)
Specifications
Specifications
| Antigen | ANXA3 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. |
| Gene | ANXA3 |
| Gene Alias | ANX3 |
| Gene Symbols | ANXA3 |
| Host Species | Mouse, Rabbit |
| Quantity | 1 Set |
| Show More |