missing translation for 'onlineSavingsMsg'
Learn More

ALDH Mouse anti-Human, Unlabeled, Clone: 44, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611194

 View more versions of this product

Product Code. 15895829

  • ONLINE EXCLUSIVE PRICE
    188.00€
    187.89€ / 1 set
    Save 0.11€
    0.06% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Aldehyde dehydrogenase (ALDH) is a ubiquitous enzyme located in nearly all mammalian tissues. It catalyzes the irreversible oxidation of a range of aliphatic and aromatic aldehydes to their corresponding carboxylic acids. There are multiple isoforms of ALDH which are subdivided into three classes. Class I includes the cytosolic isoforms. Class II includes the mitochondrial isoforms. Class III includes the microsomal, cytosolic tumor specific, and cytosolic dioxin-inducible forms. At least twelve human ALDH isoforms have been identified. Mutations of many of these proteins such as ALDH1, ALDH2, ALDH4, and ALDH10 have been implicated in multiple human metabolic disorders and clinical abnormalities. At the amino acid level, human ALDH isoforms exhibit a wide range of diversity (15% to about 80%). However, multiple protein regions have been highly conserved and are important for functional activities. A well-characterized member of the human ALDH family is ALDH1. It plays a major role in the biosynthesis of retinoic acid from retinol (vitamin A). Retinoic acid, the biologically
active form of retinol, is a regulator of cellular proliferation, differentiation, and survival.

Host Species: Mouse
Clone: 44
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human ALDH1 aa. 7-128
Formula Weight [Chemical]: 55kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ALDH
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human ALDH1 aa. 7-128
50 μg
Cell Biology
Human
IgG1
Western Blot
44
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.