missing translation for 'onlineSavingsMsg'
Learn More

Anti-Albumin antibody [RM1208], Abcam

Product Code. 32072420 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
1 mL
Unit Size:
1mL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32072420 1 mL 1mL
32070481 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 32072420 Supplier Abcam Supplier No. ab3207511mL
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Polyclonal Antibody

Rabbit Recombinant Multiclonal Albumin antibody. Suitable for IHC-P, WB, ICC/IF and reacts with Human, Mouse, Rat samples.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen ALB (Albumin)
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Western Blot
Cellular Localization Secreted
Classification Recombinant Polyclonal
Clone RM1208
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 40% Glycerol (glycerin, glycerine), 0.05% BSA
Gene ALB
Gene Accession No. P02768
Host Species Rabbit
Immunogen ALB (Albumin)
Purification Method Affinity purification
Quantity 1 mL
Regulatory Status RUO
Research Discipline Metabolism
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Blue Ice
Target Gene ALB
Target Molecule ALB (Albumin)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.