Learn More
anti-AK1 Ab-1 Matched Antibody Pair, Abnova™
Product Code. 16052818
Change view
Click to view available options
Quantity:
1 Set
Packungsgröße:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
16052818 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16052818 Lieferant Abnova Lieferanten-Nr. H00000203AP11.1Set
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beinhaltet
Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-AK1 (100μg; Detection antibody: mouse monoclonal anti-AK1, IgG1 Kappa (20μg; Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Host Species: Rabbit, Mouse
Isotype: IgG, IgGk1, κ
Cross Reactivity: Human
ELISA Pair
Spezifikation
Antigen AK1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human AK1.
Gene AK1
Gene Symbols AK1
Host Species Mouse, Rabbit
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 203
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-AK1 (100 μg) 2. Detection antibody: mouse monoclonal anti-AK1, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG
Mehr anzeigen Weniger anzeigen