Learn More
AIP1 Mouse, Unlabeled, Clone: 49, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611621
Description
Apoptosis is a selective process of genetically programmed cell death which occurs during normal cell differentiation and development of multicellular organisms. In vertebrates, T cell and neuronal development are probably the best characterized systems for the study of apoptosis. ALG-2 and ALG-3 (apoptosis-linked genes 2 and 3) were identified as low molecular weight Ca2+ -binding proteins essential for apoptosis through the activation of the Fas receptor in T cells. ALG-2 Interacting Protein 1 (AIP1/Alix) is a ubiquitous protein that associates with ALG-2 in the cytosol in a Ca2+; dependent manner. AIP1 is homologous to the yeast protein, BRO1, which has been implicated in Pkc1p- AP kinase signaling. A truncated form of AIP1 protects against serum starvation-, etoposide-, and staurosporine-induced cell death. In addition, the C-terminal proline rich region of AIP1 facilitates interaction with SH3 domain-containing protein expressed in tumorigenic astrocytes (SETA) and this interaction may be important for mediating DNA damage-dependent apoptosis in astrocytes. Thus, AIP1 interacts with ALG-2 or SETA, or both, during activation of cell death pathways in a variety of cell types.
Host Species: Mouse
Clone: 49
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Mouse AIP1 aa. 375-580
Immunofluorescence, Western Blotting
Specifications
| AIP1 | |
| Monoclonal | |
| 250μg/mL | |
| ALG-2 Interacting Protein 1; Alix | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 49 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Mouse AIP1 aa. 375-580 | |
| 150 μg | |
| Cell Biology | |
| Mouse, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.