missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A8/ANXA8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | Annexin A8/ANXA8 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18696752
|
Novus Biologicals
NBP2-97052-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669894
|
Novus Biologicals
NBP2-97052-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Annexin A8/ANXA8 Polyclonal antibody specifically detects Annexin A8/ANXA8 in Human samples. It is validated for Western Blot, ELISASpecifications
| Annexin A8/ANXA8 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 653145 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| annexin A8, Annexin VIII, Annexin-8, ANX8, FLJ53095, VAC-beta, Vascular anticoagulant-beta | |
| Produced in rabbits immunized with purified, recombinant Human Annexin A8/ANXA8 (Accession#: AAH73755.1; Met1-Pro327). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title