missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A4 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Specifications
| Antigen | Annexin A4 |
|---|---|
| Concentration | 650 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234859
|
Proteintech
29508-1-AP-20UL |
20 μL |
145.00€ 129.00€ / 20µL Save 16.00€ 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30237100
|
Proteintech
29508-1-AP-150UL |
150 μL |
410.00€ 349.00€ / 150µL Save 61.00€ 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Annexin A4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Annexin A4. This antibody recognizes human, rat, mouse antigen. The Annexin A4 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Annexin A4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| ANX4; DKFZp686H02120; MGC75105; PIG28; ZAP36 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 650 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 307 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title