missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Antibody (SAIC-13B-19) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
244.00€ - 635.00€
Specifications
| Antigen | Annexin A1 |
|---|---|
| Clone | SAIC-13B-19 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649553
|
Novus Biologicals
NBP3-20092-0.025mg |
0.025 mg |
244.00€
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625574
|
Novus Biologicals
NBP3-20092-0.2mg |
0.2 mg |
635.00€
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Mass SpectrometrySpecifications
| Annexin A1 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Peptide 'AAYLQETGKPLDETLK' derived from Annexin A1 conjugated to KLH. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| SAIC-13B-19 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Mass Spectrometry | |
| Unconjugated | |
| Rabbit | |
| Protein Kinase, Signal Transduction | |
| PBS | |
| 301 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title