Learn More
ANKRD46 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35871-20ul
This item is not returnable.
View return policy
Description
ANKRD46 Polyclonal antibody specifically detects ANKRD46 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| ANKRD46 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| ankyrin repeat domain 46 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-232 of human ANKRD46 (NP_001257308.1).,, Sequence:, MSYVFVNDSSQTNVPLLQACIDGDFNYSKRLLESGFDPNIRDSRGRTGLHLAAARGNVDICQLLHKFGADLLATDYQGNTALHLCGHVDTIQFLVSNGLKIDICNHQGATPLVLAKRRGVNKDVIRLLESLEEQEVKGFNRGTHSKLETMQTAESESAMESHSLLNPNLQQGEGVLSSFRTTWQEFVEDLGFWRVLLLIFVIALLSLGIAYYRRTLRLGSFARQDRSRIQAI | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 157567 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |